Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40512

Supelco

3,4-Dimethylhexane

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$78.20

$78.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$78.20

About This Item

Linear Formula:
CH3CH2CH(CH3)CH(CH3)CH2CH3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1718744
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$78.20


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.403 (lit.)
n20/D 1.405

bp

119 °C (lit.)

density

0.72 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCC(C)C(C)CC

InChI

1S/C8H18/c1-5-7(3)8(4)6-2/h7-8H,5-6H2,1-4H3

InChI key

RNTWWGNZUXGTAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service