Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40426

Sigma-Aldrich

(Chloromethyl)polystyrene

extent of labeling: ~3.4 mmol/g Cl loading

Synonym(s):

Merrifield resin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

extent of labeling

~3.4 mmol/g Cl loading

matrix

(crosslinked with 2% DVB)

particle size

200-400 mesh

InChI

1S/CH3Cl/c1-2/h1H3

InChI key

NEHMKBQYUWJMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service