Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

40262

Sigma-Aldrich

N,N-Dimethylformamide dibutyl acetal

for esterification of fatty acids, ≥98.0%

Synonym(s):

1,1-Dibutoxy-N,N-dimethylmethylamine, 1,1-Dibutoxytrimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH(OCH2CH2CH2CH3)2
CAS Number:
Molecular Weight:
203.32
Beilstein:
1098693
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

quality

for esterification of fatty acids

refractive index

n20/D 1.417

bp

93 °C/12 mmHg (lit.)

density

0.853 g/mL at 20 °C (lit.)

SMILES string

CCCCOC(OCCCC)N(C)C

InChI

1S/C11H25NO2/c1-5-7-9-13-11(12(3)4)14-10-8-6-2/h11H,5-10H2,1-4H3

InChI key

GSFFXKGTGPMVLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylformamide dibutyl acetal (1,1-Dibutoxytrimethylamine, 1,1-Dibutoxy-N,N-dimethylmethylamine) is a dimethylformamide dialkyl acetal.[1] Its synthesis has been reported by Meerwin and coworkers by reaction of N,N-Dimethylformamide diethyl acetal with butanol.[2]

Application

N,N-Dimethylformamide dibutyl acetal is the suitable reagent used for the simultaneous quantification of cocaine and its primary metabolite, benzoyl ecgonine, from a urine matrix by gas-chromatography.[1] It may be used as reagent for the esterification of fatty acids.[3] [4]

Other Notes

Reagent for the esterification of fatty acids[3]; Review[4]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.P. Thenot et al.
Analytical Letters, 217-217 (1972)
DeWolfe RH.
Carboxylic Ortho Acid Derivatives: Preparation and Synthetic Applications (Academic press), 14, 424-424 (2012)
N C Jain et al.
Journal of forensic sciences, 22(1), 7-16 (1977-01-01)
A gas chromatographic procedure has been developed for the simultaneous determination of cocaine and benzoyl ecgonine in urine specimens. The two drugs are extracted by isopropanol/chloroform from urine samples saturated with a bisalt buffer. The organic extract is evaporated to
R.F. Abdulla et al.
Tetrahedron, 35, 1675-1675 (1979)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service