Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

39863

Supelco

(±)-Sotalol hydrochloride

analytical standard

Synonym(s):

N-[4-[1-Hydroxy-2-(isopropylamino)ethyl]phenyl]methanesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20N2O3S · HCl
CAS Number:
Molecular Weight:
308.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤2.0% water

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

Cl.CC(C)NCC(O)c1ccc(NS(C)(=O)=O)cc1

InChI

1S/C12H20N2O3S.ClH/c1-9(2)13-8-12(15)10-4-6-11(7-5-10)14-18(3,16)17;/h4-7,9,12-15H,8H2,1-3H3;1H

InChI key

VIDRYROWYFWGSY-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Van Asbroeck et al.
Acta clinica Belgica, 69(2), 132-134 (2014-04-15)
Acquired long-QT syndrome is an iatrogenic disorder, usually induced by drugs, which can cause life-threatening arrhythmias. We present a case report on an acquired long-QT syndrome with an interesting confluence of circumstances, and comment on aetiology and treatment.
Nancy M Allen LaPointe et al.
The American journal of cardiology, 115(3), 316-322 (2014-12-11)
Little is known in clinical practice about antiarrhythmic drug (AAD) use in patients with atrial fibrillation (AF) (particularly younger ones) who do not have structural heart disease. Using the MarketScan database, we identified patients <65 years without known coronary artery disease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service