Skip to Content
MilliporeSigma
All Photos(3)

Documents

39851

Millipore

HybriScanI Alicyclobacillus

suitable for microbiology

Synonym(s):

Alicyclobacillus HybriScan I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116131
NACRES:
NB.11

shelf life

limited shelf life, expiry date on the label

Quality Level

application(s)

microbiology

storage temp.

2-8°C

General description

HybriScan I kits are intended for the identification and result confirmation of microorganisms using rRNA as the detection target. Several customers also use it to detect microorganisms in food, beverage and water samples. The principle is based on in situ sandwich hybridization with two specific probes. As the rRNA of dead cells is degraded in the presence of RNase, only living cells are detected. Each cell has several thousand rRNA copies, so neither PCR nor a polymerase is needed for this technology, which means that this molecular biology test is not inhibited by the sample matrix. We recommend a 72±4-hour enrichment in BAT broth (Cat. No. 1.07993)

Application

The rapid and innovative test system for the identification of Alicyclobacillus.

Features and Benefits

  • Sensitivity (cfu/assay): 1000

Legal Information

HybriScan is a trademark of ScanBec GmbH

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1A - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Alicyclobacillus are acidophilic, strictly aerobic, and spore-forming spoilage bacteria that can cause increased turbidity, color changes and unpleasant sensory properties in juices. Testing is performed according to IFU method No 12 (2019).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service