Skip to Content
MilliporeSigma
All Photos(1)

Documents

393363

Sigma-Aldrich

4-Pentylbicyclo[2.2.2]octane-1-carboxylic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24O2
CAS Number:
Molecular Weight:
224.34
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

99%

form

powder or crystals

mp

160-162 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCCCC[C@@]12CC[C@@](CC1)(CC2)C(O)=O

InChI

1S/C14H24O2/c1-2-3-4-5-13-6-9-14(10-7-13,11-8-13)12(15)16/h2-11H2,1H3,(H,15,16)/t13-,14-

InChI key

IAQHPZFALQQESM-HDJSIYSDSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Wilde et al.
Journal of chromatography. A, 1378, 74-87 (2015-01-03)
Although bicyclic acids have been reported to be the major naphthenic acids in oil sands process-affected water (OSPW) and a well-accepted screening assay indicated that some bicyclics were the most acutely toxic acids tested, none have yet been identified. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service