Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38828

Sigma-Aldrich

6,7-Dimethoxy-4-(trifluoromethyl)coumarin

for fluorescence, ≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9F3O4
CAS Number:
Molecular Weight:
274.19
Beilstein:
8327618
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

Assay

≥98.0% (TLC)

mp

158-159 °C (lit.)

solubility

DMF: soluble
DMSO: soluble

fluorescence

λex 358 nm; λem 490 nm in DMSO

SMILES string

COc1cc2OC(=O)C=C(c2cc1OC)C(F)(F)F


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service