Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38462

Supelco

Rebaudioside A

analytical standard

Synonym(s):

(4α)-13-[(2-O-β-D-glucopyranosyl-3-O-β-D­glucopyranosyl-β-D-glucopyranosyl)-oxy]kaur-6-en-8-oic acid β-D­glucopyranosyl ester, Glycoside A3, Glycoside X, Stevioside a3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H70O23
CAS Number:
Molecular Weight:
967.01
Beilstein:
6470556
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤12.0% water

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C[C@@]12CCC[C@](C)([C@H]1CC[C@]34CC(=C)C(CC[C@@H]23)(C4)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)[C@H]5O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)C(=O)O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O

InChI

1S/C44H70O23/c1-17-11-43-9-5-22-41(2,7-4-8-42(22,3)40(59)66-38-33(58)30(55)26(51)20(14-47)62-38)23(43)6-10-44(17,16-43)67-39-35(65-37-32(57)29(54)25(50)19(13-46)61-37)34(27(52)21(15-48)63-39)64-36-31(56)28(53)24(49)18(12-45)60-36/h18-39,45-58H,1,4-16H2,2-3H3/t18-,19-,20-,21-,22+,23+,24-,25-,26-,27-,28+,29+,30+,31-,32-,33-,34+,35-,36+,37+,38+,39+,41-,42-,43-,44+/m1/s1

InChI key

HELXLJCILKEWJH-NCGAPWICSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Componentof stevia.
Rebaudioside A is a type of naturally occurring steviol glycoside, which can be obtained from Stevia rebaudiana species of plants.[1][2] It finds applications as food and beverage sweetener.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Overview: the history, technical function and safety of rebaudioside A, a naturally occurring steviol glycoside, for use in food and beverages
Carakostas.C.M, et al.
Food And Chemical Toxicology, 46, S1-S10 (2008)
The hemodynamic effects of rebaudioside A in healthy adults with normal and low-normal blood pressure
Maki.C.K, et al.
Food And Chemical Toxicology, 46, S40-S46 (2008)
M C Carakostas et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 7, S1-S10 (2008-06-17)
Rebaudioside A is a sweet tasting steviol glycoside extracted and purified from Stevia rebaudiana (Bertoni). Steviol glycosides can currently be used as a food ingredient in only a handful of countries. Questions on specifications, safety and special population effects have
Magdalena Simlat et al.
PloS one, 15(3), e0230755-e0230755 (2020-03-29)
Melatonin (MEL) can act as a plant growth regulator and biostimulator in stressful situations. Using MEL in seed pretreatment also affects the future growth of plants. Therefore, this research investigated the effects of MEL on seed germination and seedling growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service