Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

381888

Sigma-Aldrich

2,3-Naphthalenedicarbonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CN)2
CAS Number:
Molecular Weight:
178.19
Beilstein:
2574929
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

97%

form

powder

mp

253-255 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

N#Cc1cc2ccccc2cc1C#N

InChI

1S/C12H6N2/c13-7-11-5-9-3-1-2-4-10(9)6-12(11)8-14/h1-6H

InChI key

KNBYJRSSFXTESR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to a new molecular semiconductor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Chemical Communications, 442-442 (1991)
Maxim A Faraonov et al.
Dalton transactions (Cambridge, England : 2003), 46(11), 3547-3555 (2017-02-28)
Crystalline anionic salts of titanyl macrocycles with acceptor substituents or an extended π-system have been obtained for the first time: (PPN

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service