Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37212

Supelco

Ammonium test kit Quantofix®

10-400 mg/L (ammonium)

Synonym(s):

Quantofix® Ammonium test kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

description

semi-quantitative

specific analyte(s)

ammonium

measuring range

10-400 mg/L (ammonium)

application(s)

agriculture
environmental
food and beverages
general analytical

detection method

colorimetric

General description

Contents:
Kit including reagents and test sticks for 100 tests.

Storage:
Avoid exposing the sticks to sunlight and moisture. Store the kit below +30 °C in a dry place.

Gradation:
0 - 10 - 25 - 50 - 100 - 200 - 400 mg/l NH4+

Application

It was used in testing ammonia concentration of Serratia odorifera and also in establishing a calibration curve with 0.5-50 μmol concentrations of ammonia; in an attempt to analyse volatile emission and understand biological impact of volatile compounds on Arabidopsis thaliana.

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marco Kai et al.
Applied microbiology and biotechnology, 88(4), 965-976 (2010-08-19)
Bacteria emit a wealth of volatiles. The combination of coupled gas chromatography/mass spectrometry (GC/MS) and proton-transfer-reaction mass spectrometry (PTR-MS) analyses provided a most comprehensive profile of volatiles of the rhizobacterium Serratia odorifera 4Rx13. An array of compounds, highly dominated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service