Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37210

Supelco

Phosphate test kit Quantofix®

3-100 mg/L (PO43-)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319
NACRES:
NA.21

description

semi-quantitative

specific analyte(s)

phosphate

measuring range

3-100 mg/L (PO43-)

application(s)

agriculture
environmental
food and beverages
general analytical

detection method

colorimetric

General description

QUANTOFIX® test kit is a ready to use precalibrated kit for phosphate testing in solutions.

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Barden
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 34(10), 1271-1279 (1986-10-01)
The intragranular location of carboxyl groups was tinctorially determined in human substantia nigra neuromelanin granules, human inferior olive lipofuscin granules, and mouse meningeal melanosomes. Soluble and insoluble lipid was stained with beta naphthol Sudans in unoxidized and oxidized frozen and
R J Thurston et al.
Biology of reproduction, 48(2), 393-402 (1993-02-01)
Protease and basic amidase activity was found in the seminal plasma of the domestic turkey. Amidase activity, measured through use of N-alpha-benzoyl-DL-arginine-p-nitroanilide-HCL (BAPNA), was 23-28 times greater for turkey than for guinea fowl or chicken. Within the reproductive tract, seminal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service