Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37063

Supelco

Special indicator paper

pH 5.5-9.0

Synonym(s):

Indicator paper, special

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319
NACRES:
NA.21

feature

non-bleeding: no
refill: no

pH

5.5-9.0

suitability

corresponds for shade of color acc. to scale

application(s)

environmental
food and beverages
general analytical
industrial hygiene

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide

General description

Special indicator papers cover range from 2 to 5 pH units in general. It has the capacity to change colour at particular pH value. Nonbleeding indicator papers are prepared by either direct dyes or reactive dyes. During reactive dyes procedure, pure linters are suspended in water and reactive dye is added. The mixture is made alkaline for the reaction to start. The fiber pulp is centrifuged, washed to be electrolyte-free and processed as pH paper. Paper produced in this way is bonded to plastic material in order to be used as stick or strip.[1]

Application

It may be used in volatile cyanide detection during Chitinase activity assay to understand the antifungal and phytostimulating characteristic of B. subtilis Ch-13 strain.[2]

Packaging

6 boxes with 1 color chart
roll with approx. 5 m

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antifungal and phytostimulating characteristics of Bacillus subtilis Ch-13 rhizospheric strain, producer of bioprepations.
Makarova, N. M., A. I. Shaposhnikov, and L. V. Kravchenko.
Applied Biochemistry and Microbiology, 45, 419-423 (2009)
Klaus Hunger
Industrial Dyes: Chemistry, Properties, Applications, 541-541 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service