Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37024

Supelco

Monocrotaline

analytical standard

Synonym(s):

Crotaline, Monocrotaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23NO6
CAS Number:
Molecular Weight:
325.36
Beilstein:
48732
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

204 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

[H][C@@]12[C@H]3CCN1CC=C2COC(=O)[C@](C)(O)[C@](C)(O)[C@@H](C)C(=O)O3

InChI

1S/C16H23NO6/c1-9-13(18)23-11-5-7-17-6-4-10(12(11)17)8-22-14(19)16(3,21)15(9,2)20/h4,9,11-12,20-21H,5-8H2,1-3H3/t9-,11+,12+,15+,16-/m0/s1

InChI key

QVCMHGGNRFRMAD-XFGHUUIASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Monocrotaline is a pyrrolizidine alkaloid that is found in plants, which belongs to the genus Crotalaria sp.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Reconstitution

Dried down, 100 μg/mL after reconstitution with 1 mL of water

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward A Pankey et al.
American journal of physiology. Heart and circulatory physiology, 305(9), H1288-H1296 (2013-09-03)
Cardiovascular responses to the tyrosine kinase inhibitor imatinib were investigated in the rat. Intravenous injections of 0.3-30 mg/kg imatinib produced small decreases in pulmonary arterial pressure, larger dose-dependent decreases in systemic arterial pressure, and no change or small increases in
Satoshi Matsuo et al.
The Journal of thoracic and cardiovascular surgery, 149(3), 902-909 (2014-12-03)
Lung ischemia-reperfusion (IR) injury during cardiopulmonary surgery is associated with postoperative morbidity and mortality, particularly in patients with pulmonary hypertension (PH). Using a rat model for monocrotaline-induced PH, we investigated the protective effect of rosuvastatin against IR injury in lungs
D W Wilson et al.
Critical reviews in toxicology, 22(5-6), 307-325 (1992-01-01)
Monocrotaline (MCT) is an 11-membered macrocyclic pyrrolizidine alkaloid (PA) that causes a pulmonary vascular syndrome in rats characterized by proliferative pulmonary vasculitis, pulmonary hypertension, and cor pulmonale. Current hypotheses of the pathogenesis of MCT-induced pneumotoxicity suggest that MCT is activated
Emmy Manders et al.
American journal of physiology. Lung cellular and molecular physiology, 303(12), L1070-L1078 (2012-09-11)
We previously demonstrated that diaphragm muscle weakness is present in experimental pulmonary arterial hypertension (PH). However, the nature of this diaphragm weakness is still unknown. Therefore, the aim of this study was to investigate whether changes at the sarcomeric level
Jose G Gomez-Arroyo et al.
American journal of physiology. Lung cellular and molecular physiology, 302(4), L363-L369 (2011-10-04)
Severe forms of pulmonary arterial hypertension (PAH) are characterized by various degrees of remodeling of the pulmonary arterial vessels, which increases the pulmonary vascular resistance and right ventricular afterload, thus contributing to the development of right ventricle dysfunction and failure.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service