Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36725

Sigma-Aldrich

Dicyclopropylmethanol

purum, ≥98.5% (GC)

Synonym(s):

DCPM-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O
CAS Number:
Molecular Weight:
112.17
Beilstein:
2203215
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

purum

Assay

≥98.5% (GC)

refractive index

n20/D 1.464

bp

69 °C/13 mmHg (lit.)

density

0.951 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(C1CC1)C2CC2

Other Notes

Protecting group reagent for carboxylic acids; the formed esters are very mildly and selectively cleaved[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L.A. Carpino et al.
The Journal of Organic Chemistry, 60, 7718-7718 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service