Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36617

Sigma-Aldrich

(1R,2R)-1,2-Dicyclohexyl-1,2-ethanediol

purum, ≥97.0% (GC)

Synonym(s):

(1R,2R)-1,2-Dicyclohexylethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H11CH(OH)-]2
CAS Number:
Molecular Weight:
226.36
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

optical activity

[α]20/D −2.3±0.3°, c = 1% in chloroform stab. with amylenes

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

138-142 °C (lit.)
138-142 °C

SMILES string

O[C@H](C1CCCCC1)[C@H](O)C2CCCCC2

InChI

1S/C14H26O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h11-16H,1-10H2/t13-,14-/m1/s1

InChI key

CKHLARJSOMXDKL-ZIAGYGMSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Auxiliary used in highly enantio- and diastereoselective syntheses via boronic esters

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.S. Matteson et al.
The Journal of Organic Chemistry, 58, 6545-6545 (1993)
D.S. Matteson et al.
Journal of the American Chemical Society, 112, 3964-3964 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service