Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36517

Supelco

Flurochloridon

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10Cl2F3NO
CAS Number:
Molecular Weight:
312.12
Beilstein:
1544703
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

description

mixture of isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

FC(F)(F)c1cccc(c1)N2CC(CCl)C(Cl)C2=O

InChI

1S/C12H10Cl2F3NO/c13-5-7-6-18(11(19)10(7)14)9-3-1-2-8(4-9)12(15,16)17/h1-4,7,10H,5-6H2

InChI key

OQZCSNDVOWYALR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Industrial hygiene in the use of the herbicide Raiser in cotton growing].
V V Tarasov et al.
Gigiena truda i professional'nye zabolevaniia, (4)(4), 9-10 (1992-01-01)
Rebeca Sultana et al.
Journal of infection in developing countries, 6(2), 156-165 (2012-02-18)
Highly pathogenic avian influenza (H5N1) virus (known as "bird flu") is an important public health concern due to its potential to infect humans and cause a human pandemic. Bangladesh is a high-risk country for an influenza pandemic because of its
Noelia Nikoloff et al.
Toxicology in vitro : an international journal published in association with BIBRA, 26(1), 157-163 (2011-11-15)
The in vitro effects of flurochloridone (FLC) and its formulations Twin Gold Pack® (25% a.i.) and Rainbow® (25% a.i.) were evaluated on Chinese hamster ovary (CHO-K1) cells by genotoxicity [sister chromatid exchange (SCE)] and cytotoxicity [cell-cycle progression, proliferative rate index
Armagan Kaya et al.
Ecotoxicology and environmental safety, 106, 232-238 (2014-05-27)
In this study, we comparatively evaluated the effects of the flurochloridone as well as flurochloridone and exogenously applied salicylic acid (SA) on Helianthus annuus L. to find out herbicide-induced toxicity reducing influence of SA. We examined and compared the physiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service