Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

36236

Supelco

(−)-Curcumol

analytical standard

Synonym(s):

(3S,3aS,5S,6R,8aS)-Octahydro-3-methyl-8-methylene-5-(1-methylethyl)-6H-3a,6-epoxyazulen-6-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O2
CAS Number:
Molecular Weight:
236.35
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC(C)[C@@H]1C[C@@]23O[C@]1(O)CC(=C)[C@@H]2CC[C@@H]3C

InChI

1S/C15H24O2/c1-9(2)13-8-14-11(4)5-6-12(14)10(3)7-15(13,16)17-14/h9,11-13,16H,3,5-8H2,1-2,4H3/t11-,12-,13-,14-,15+/m0/s1

InChI key

QRMPRVXWPCLVNI-YYFQZIEXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S Zhang et al.
Journal of pharmaceutical and biomedical analysis, 48(3), 1024-1028 (2008-08-30)
The rhizomes of Curcuma phaeocaulis, Curcuma kwangsiensis, Curcuma wenyujin and Curcuma longa are used as Ezhu or Jianghuang in traditional Chinese medicine for a long time. Due to their similar morphological characters, it is difficult to distinguish their origins of
K X Huang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 25(3), 163-165 (2002-09-06)
To study the effective ingredients of Curcuma aromatica. Solvent extraction was used. The constituents were isolated with resin D-101 silica gel column and thin-layer chromatography, and the structures were elucidated by physico-chemical properties and spectral analysis. Curdione, neocurdione, curcumol, tetramethylpyrazine
Xiao Zhao et al.
Biomedical chromatography : BMC, 24(4), 347-350 (2009-08-04)
A simple and sensitive capillary gas chromatography with a hydrogen flame ionization detector (GC-FID) method was developed for the determination of curcumol in rat plasma. From a variety of compounds and solvents tested, buagafuran was selected as the internal standard
Hui Zhang et al.
Chemical & pharmaceutical bulletin, 55(3), 451-454 (2007-03-03)
Further study on the microbial transformation of curcumol (1) by Cunninghamella blakesleana (AS3.970) led to the isolation of four novel metabolites. Their structures were elucidated as 3beta-hydroxy curcumol (2), 12-hydroxy curcumol (3), 1alpha-hydroxy-10beta,14-epoxy curcumol (4) and (2S,4S,5S,7S)-10-hydroxymethyl-7-isopropyl-2-methoxy-4-methyl-1-oxaspiro[4,6]undec-10-en-8-one (5) on the
Rui Zhang et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 42(9), 973-977 (2007-12-07)
To establish a high performance liquid chromatography (HPLC) coupled with tandem mass spectrometry quantitative detection method for the determination of curcumol, the main ingredient of zedoary turmeric oil fat emulsion, and investigate its pharmacokinetics in Beagle dogs, nine healthy Beagle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service