Skip to Content
MilliporeSigma
All Photos(1)

Documents

36218

Sigma-Aldrich

2,4-Dichlorophenylhydrazine hydrochloride

purum, ≥95.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NHNH2·HCl
CAS Number:
Molecular Weight:
213.49
Beilstein:
3708828
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥95.0% (AT)

impurities

~5% water

mp

220-224 °C (lit.)
220-224 °C

SMILES string

Cl.NNc1ccc(Cl)cc1Cl

InChI

1S/C6H6Cl2N2.ClH/c7-4-1-2-6(10-9)5(8)3-4;/h1-3,10H,9H2;1H

InChI key

DDWYGJVFURAIJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service