Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

35828

Supelco

2,4,5-Trichloroaniline

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2NH2
CAS Number:
Molecular Weight:
196.46
Beilstein:
879091
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

270 °C (lit.)

mp

93-95 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C6H4Cl3N/c7-3-1-5(9)6(10)2-4(3)8/h1-2H,10H2

InChI key

GUMCAKKKNKYFEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,5-Trichloroaniline is a substituted aniline with a strong activating –NH2 substituent and three weakly deactivating Cl. It is used as an intermediate for dyestuffs, pigments, pesticides, and other organic chemicals.

Application

The 2,4,5-trichloroaniline may find the following uses:

  • Experimental and theoretical study on the molecular structure and vibrational spectra of 2,4,5-trichloroaniline using Fourier transform infrared spectroscopy (FT-IR) and Fourier transform-Raman spectroscopy
  • Simultaneous reversed-phase high-performance liquid chromatographic separation of mono-, di-and trichloroanilines through an optimized gradient elution experimental design
  • Exploring aroyl/heteroaroyl iminothiazolines featuring 2,4,5-trichlorophenyl moiety as a new class of potent, selective, and in- vitro efficacious glucosidase inhibitors
  • Antimicrobial studies of some novel propenones bearing aryl furan moiety
  • Calorimetric and spectroscopic studies of complex formation of 2,4,5-Trichloroaniline with proton acceptors in carbon tetrachloride

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Liver

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Gennaro et al.
Journal of chromatography. A, 945(1-2), 287-292 (2002-02-28)
A new RP-HPLC method for the simultaneous determination of the 13 mono-, di- and trichloroanilines has been developed. In order to obtain the analyte resolution within an acceptable analysis time, a gradient elution program has been optimised through the use
Madiha Kazmi et al.
Bioorganic chemistry, 74, 134-144 (2017-08-07)
A series of iminothiazolines (4a-j) featuring 2,4,5-trichlorophenyl moiety and aroyl/heteroaroyl substituents has been prepared from readily accessible thioureas. In-vitro screening against glucosidase enzymes showed highly specific inhibition of α-glucosidase with a marked dependence of the potency upon the nature of
M Govindarajan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 231-245 (2012-07-07)
In this work, the experimental and theoretical study on the molecular structure and vibrational spectra of 2,4,5-trichloroaniline (C(6)H(4)NCl(3), abbreviated as 2,4,5-TClA) were studied. The FT-IR and FT-Raman spectra were recorded. The molecular geometry and vibrational frequencies in the ground state

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service