The solubility of the deuterated Closantel product should be similar to that of product no. 34093, the non-deuterated Closantel. It is soluble in DMSO and Dimethyl Formamide (purged with inert gas) at approximately 20 mg/ml. However, the compound is sparingly soluble in aqueous buffers. For maximum solubility, it is recommended to first dissolve it in DMSO and then dilute it into the aqueous buffer. In a 1:5 solution of DMSO:PBS (pH 7.2), Closantel has a solubility of approximately 0.16 mg/ml, and solutions are stable for no more than one day.
35359
Closantel-(benzoyl ring-13C6)
VETRANAL®, analytical standard
Synonym(s):
N-{5-Chloro-4-[(4-chlorophenyl)cyanomethyl]-2-methylphenyl}-2-hydroxy-3,5-diiodobenzamide-13C6
Select a Size
Select a Size
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
format
neat
mass shift
M+6
SMILES string
Cc1cc(C(C#N)c2ccc(Cl)cc2)c(Cl)cc1NC(=O)[13c]3[13cH][13c](I)[13cH][13c](I)[13c]3O
InChI
1S/C22H14Cl2I2N2O2/c1-11-6-15(17(10-27)12-2-4-13(23)5-3-12)18(24)9-20(11)28-22(30)16-7-14(25)8-19(26)21(16)29/h2-9,17,29H,1H3,(H,28,30)/i7+1,8+1,14+1,16+1,19+1,21+1
InChI key
JMPFSEBWVLAJKM-ZCTWQCQHSA-N
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 4
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the solubility of 35359 Closantel-(benzoyl ring-13C6)?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service