Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34765

Sigma-Aldrich

Di-n-butylmagnesium solution

~0.75 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]2Mg
CAS Number:
Molecular Weight:
138.53
Beilstein:
3535184
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

~0.75 M in diethyl ether

density

0.749 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

CCCC[Mg]CCCC

InChI

1S/2C4H9.Mg/c2*1-3-4-2;/h2*1,3-4H2,2H3;

InChI key

ODHFJIDDBSDWNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for preparing other symmetrical magnesium compounds R2Mg

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service