Skip to Content
MilliporeSigma
All Photos(1)

Documents

34761BC

Millipore

Assurance® GDS Salmonella Positive Control

For use with Assurance® GDS Salmonella Tq

Synonym(s):

Positive Control for PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116135
NACRES:
NA.85

product name

GDS Positive Control for Salmonella, BioControl, For use with GDS Salmonella

Quality Level

form

liquid

shelf life

12 mo.

packaging

vial of 5 mL (sufficient for 166 tests)

manufacturer/tradename

BioControl Systems 34761-250

storage condition

(Tightly closed)

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

storage temp.

2-8°C

suitability

Salmonella spp.

General description

GDS Positive Control for Salmonella is used with the GDS Salmonella Tq assay, which is an automated nucleic acid amplification system for the detection of Salmonella in food and environmental samples.

Application

GDS Positive Control for Salmonella is used for confirmation of Salmonella spp. detection in Assurance® GDS assays.

Components

One vial, 5 mL, sufficient for 166 tests

Compatibility

BioControl GDS

Testing Method

PCR

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service