Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34636

Supelco

Chromium(III) potassium sulfate solution

for COD determination according to DIN 38409-H44

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCr(SO4)2
CAS Number:
Molecular Weight:
283.22
MDL number:
UNSPSC Code:
41113319
PubChem Substance ID:

grade

for COD determination according to DIN 38409-H44

density

1.12 g/mL at 20 °C

application(s)

environmental
general analytical

SMILES string

[K+].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Cr.K.2H2O4S/c;;2*1-5(2,3)4/h;;2*(H2,1,2,3,4)/q+3;+1;;/p-4

InChI key

OIDPCXKPHYRNKH-UHFFFAOYSA-J

General description

The higher the pH more is the chromium (III) (crystal growth suppressor) adsorbed, on the surface of potassium sulfate crystal powder which is suspended in saturated solutions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption of growth suppressor chromium (III) on potassium sulfate crystals in aqueous solution.
Guzman LA
Journal of Crystal Growth, 186 (3), 438-445 (1998)
G M Lee et al.
The Histochemical journal, 17(11), 1221-1233 (1985-11-01)
Various pretreatments of metaphase spreads were examined to obtain optimal DNA labelling patterns while maintaining chromosome integrity during in situ hybridization procedures. Preparations of African green monkey (AGM) chromosomes fixed in methanol-acetic acid (CV-1 cell line) were treated by coating
Annette R Rowe et al.
Applied and environmental microbiology, 74(21), 6709-6719 (2008-09-09)
This study sought to characterize bacterial and archaeal populations in a perchloroethene- and butyrate-fed enrichment culture containing hydrogen-consuming "Dehalococcoides ethenogenes" strain 195 and a Methanospirillum hungatei strain. Phylogenetic characterization of this microbial community was done via 16S rRNA gene clone
V M Chikov et al.
Izvestiia Akademii nauk SSSR. Seriia biologicheskaia, (2)(2), 294-297 (1989-03-01)
Gallocyanine-chrome alum-stained pig embryonic kidney cells have paramagnetic properties. They move under the influence of gradient magnetic field (magnetophoresis). The velocity of magnetophoresis is proportional to the content of nucleic acids in cells. This allows to estimate the content of
O Schmitt et al.
Micron (Oxford, England : 1993), 28(3), 197-215 (1997-06-01)
The investigation of neurohistological specimens by image analysis has become an important tool in morphological neuroscience. The problems which arise during the processing of these images are non-trivial, especially if a pattern recognition of cells in the imaged tissue is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service