Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34541

Supelco

Malathion diacid-(dimethyl-d6)

PESTANAL®, analytical standard

Synonym(s):

2-{[Di(methoxy-d3)phosphorothioyl]sulfanyl}succinic acid, O,O-Di(methyl-d3) S-(1,2-dicarboxyethyl) phosphorodithioate, Malathion diacid-d6, Malathiondicarboxylic acid-(dimethyl-d6)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D6H5O6PS2
CAS Number:
Molecular Weight:
280.29
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

General description

Malathion diacid-(dimethyl-d6) is a deuterated analog of malathion diacid, the carboxyesterase derivative of organophosphorus insecticide malathion, wherein the O-methyl protons are substituted by deuterium.[1]

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The metabolism and synergism of malathion in resistant and susceptible strains of Tribolium castaneum (Herbst)(Coleoptera, Tenebrionidae)
Dyte CE and Rowlands DG
Journal of Stored Products Research, 4(2), 157-173 (1968)

Articles

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service