Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34503

Supelco

Secbumeton-(ethyl-d5)

PESTANAL®, analytical standard

Synonym(s):

2-(sec-Butylamino)-4-(ethyl-d5-amino)-6-methoxy-1,3,5-triazine, N2-(Ethyl-d5)-6-methoxy-N4-(1-methylpropyl)-1,3,5-triazine-2,4-diamine, Secbumeton-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D5H14N5O
CAS Number:
Molecular Weight:
230.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

CC(CC)NC1=NC(NC([2H])([2H])C([2H])([2H])[2H])=NC(OC)=N1

InChI

1S/C10H19N5O/c1-5-7(3)12-9-13-8(11-6-2)14-10(15-9)16-4/h7H,5-6H2,1-4H3,(H2,11,12,13,14,15)/i2D3,6D2

InChI key

ZJMZZNVGNSWOOM-QKLSXCJMSA-N

General description

Secbumeton-(ethyl-d5) is an isotope labeled analog of the triazine herbicide secbumeton, wherein the N-ethyl protons are replaced by deuterium.

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis, potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of triazine herbicides in vegetables by ionic liquid foam floatation solid phase extraction high performance liquid chromatography
Zhang L, et al.
Journal of Chromatography. B, Biomedical Applications, 953, 132-137 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service