Skip to Content
MilliporeSigma
All Photos(2)

Documents

34216

Supelco

Ipronidazole-d3

VETRANAL®, analytical standard

Synonym(s):

2-Isopropyl-1-methyl-d3-5-nitro-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7D3H8N3O2
CAS Number:
Molecular Weight:
172.20
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])n1c(ncc1[N+]([O-])=O)C(C)C

InChI

1S/C7H11N3O2/c1-5(2)7-8-4-6(9(7)3)10(11)12/h4-5H,1-3H3/i3D3

InChI key

NTAFJUSDNOSFFY-HPRDVNIFSA-N

Application

Ipronidazole-d3 may be used as an analytical reference standard for the determination of the analyte in animal tissues by liquid chromatography tandem mass spectrometry (LC-MS/MS) and ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) using a QUick, Easy, CHeap, Effective, Rugged and Safe (QuEChERS) approach.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue method for identification and quantification of avermectins, benzimidazoles and nitroimidazoles residues in bovine muscle tissue by ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) using a QuEChERS approach.
da Silva GR, et al.
Talanta, 171(1-2), 307-320 (2017)
The development and validation of a multiclass liquid chromatography tandem mass spectrometry (LC-MS/MS) procedure for the determination of veterinary drug residues in animal tissue using a QuEChERS (QUick, Easy, CHeap, Effective, Rugged and Safe) approach.
Stubbings, George and Bigwood, Timothy
Analytica Chimica Acta, 637(1-2), 68-78 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service