Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34183

Supelco

(S)-3-Anilino-5-methyl-5-phenylimidazolidine-2,4-dione

PESTANAL®, analytical standard

Synonym(s):

(5S)-5-Methyl-5-phenyl-3-(phenylamino)-2,4-imidazolidinedione, Fenamidone Metabolite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3O2
CAS Number:
Molecular Weight:
281.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C[C@]1(NC(=O)N(Nc2ccccc2)C1=O)c3ccccc3

InChI

1S/C16H15N3O2/c1-16(12-8-4-2-5-9-12)14(20)19(15(21)17-16)18-13-10-6-3-7-11-13/h2-11,18H,1H3,(H,17,21)/t16-/m0/s1

InChI key

KTUBSXMBPVHJQY-INIZCTEOSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosalía López-Ruiz et al.
Chemosphere, 226, 36-46 (2019-03-27)
In this study, fenamidone, propamocarb and their transformation products were monitored in cherry tomato, cucumber, and courgette samples. A mixture of both compounds, which have different physico-chemical characteristics, are commercially available (Consento®). For analysis, ultra high-performance liquid chromatography coupled to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service