Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34105

Supelco

PCB No 149

analytical standard

Synonym(s):

2,2′,3,4′,5′,6-Hexachlorobiphenyl, 2,2′,3,4′,5′,6-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Cl6
CAS Number:
Molecular Weight:
360.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1ccc(Cl)c(c1Cl)-c2cc(Cl)c(Cl)cc2Cl

InChI

1S/C12H4Cl6/c13-6-1-2-7(14)12(18)11(6)5-3-9(16)10(17)4-8(5)15/h1-4H

InChI key

LKHLFUVHHXCNJH-UHFFFAOYSA-N

General description

PCB No 149 solution belongs to the class of polychlorinated biphenyls (PCBs), a group of highly toxic pollutants commonly found in various environmental matrices and food products. They find a variety of applications as flame retardants, plasticizers, dielectric and heat-transfer fluids, etc.

Application

PCB No 149 solution may be used as an analytical reference standard for the determination of the analyte in air samples and wastewater samples using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous analysis of organochlorine pesticides and polychlorinated biphenyls in air samples by using accelerated solvent extraction (ASE) and solid-phase micro-extraction (SPME) coupled to gas chromatography dual electron capture detection
Mokbel H, et al.
Environmental Science and Pollution Research International, 23(8), 8053-8063 (2016)
Solid-phase extraction combined with large volume injection-programmable temperature vaporization?gas chromatography?mass spectrometry for the multiresidue determination of priority and emerging organic pollutants in wastewater
Bizkarguenaga E, et al.
Journal of Chromatography A, 1247, 104-117 (2012)
Isolation and characterisation of polychlorinated biphenyl (PCB) degrading fungi from a historically contaminated soil
Tigini V, et al.
Microbial cell factories, 8(1), 5-5 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service