Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34085

Supelco

Ethychlozate

PESTANAL®, analytical standard

Synonym(s):

Ethyl 5-chloro-1H-indazole-3-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11ClN2O2
CAS Number:
Molecular Weight:
238.67
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)Cc1n[nH]c2ccc(Cl)cc12

InChI

1S/C11H11ClN2O2/c1-2-16-11(15)6-10-8-5-7(12)3-4-9(8)13-14-10/h3-5H,2,6H2,1H3,(H,13,14)

InChI key

GLPZEHFBLBYFHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethychlozate is a highly functional compound, commonly used as a plant growth regulator of synthetic auxin for agricultural commodities.

Application

Ethychlozate may be used as an analytical reference standard for the estimation of the analyte in fruits and vegetables using QuEChERS (quick, easy, cheap, effective, rugged and safe) method with liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Establishment of Analytical Method for Residues of Ethychlozate, a Plant Growth Regulator, in Brown Rice, Mandarin, Pepper, Potato, and Soybean Using HPLC/FLD.
Kim YJ, et al.
Korean Journal of Environmental Agriculture, 34(2), 111-119 (2015)
Simultaneous determination of 10 plant growth promoters in fruits and vegetables with a modified QuEChERS based liquid chromatography tandem mass spectrometry method.
Shuiying R, et al.
Analytical Methods : Advancing Methods and Applications, 7(21), 9130-9136 (2015)
Satoshi Takatsuki et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 43(1), 30-34 (2002-05-10)
A method was developed for the analysis of ethychlozate (CIE) and its decomposition compound, 5-chloro-3(1H)-indazolylacetic acid (CIA) in fruits by HPLC and LC/MS. The sample was homogenized with 1 mol/L HC1, and CIE and CIA were extracted with 5 mol/L

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service