Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

34006

Supelco

1-Amino-2,4-imidazolidinedione-13C3

VETRANAL®, analytical standard

Synonym(s):

AHD-13C3

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,140.00

$1,140.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$1,140.00

About This Item

Empirical Formula (Hill Notation):
13C3H5N3O2
CAS Number:
Molecular Weight:
118.07
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$1,140.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

mass shift

M+3

SMILES string

NN1[13CH2][13C](=O)N[13C]1=O

InChI

1S/C3H5N3O2/c4-6-1-2(7)5-3(6)8/h1,4H2,(H,5,7,8)/i1+1,2+1,3+1

InChI key

KVYKDNGUEZRPGJ-VMIGTVKRSA-N

Application

1-Amino-2,4-imidazolidinedione-13C3 may be used as an internal standard for the quantification of 1-amino-2,4-imidazolidinedione in shrimp using liquid chromatography with tandem mass spectrometry (LC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of nitrofuran metabolites and chloramphenicol in shrimp with a single extraction and LC-MS/MS analysis.
El-Demerdash A, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 98(3), 595-601 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service