Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

33887

Supelco

Desmethyl-formamido-pirimicarb

PESTANAL®, analytical standard

Synonym(s):

5,6-Dimethyl-2-(N-formylmethylamino)-4-pyrimidinyl dimethylcarbamate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$223.00

$223.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$223.00

About This Item

Empirical Formula (Hill Notation):
C11H16N4O3
CAS Number:
Molecular Weight:
252.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$223.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CN(C)C(=O)Oc1nc(nc(C)c1C)N(C)C=O

InChI

1S/C11H16N4O3/c1-7-8(2)12-10(15(5)6-16)13-9(7)18-11(17)14(3)4/h6H,1-5H3

InChI key

GDEAMEURJBBCOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Desmethyl-formamido-pirimicarb is a derivative of the pesticide, pirimicarb.[1]

Application

Desmethyl-formamido-pirimicarb may be used as an analytical reference standard for the determination of the analyte in food samples using modified quick, easy, cheap, effective, rugged, and safe (QuEChERS) extraction method followed by liquid chromatography-tandem mass spectrometry (LC-MS/MS).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

evaluation of some pesticide residues in food (1976)
Validation and use of a fast sample preparation method and liquid chromatography-tandem mass spectrometry in analysis of ultra-trace levels of 98 organophosphorus pesticide and carbamate residues in a total diet study involving diversified food types.
Chung SWC and Chan BTP
Journal of Chromatography A, 1217(29), 4815-4824 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service