Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

337897

Sigma-Aldrich

Mercury(II) acetate

97%

Synonym(s):

Mercuric acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Hg
CAS Number:
Molecular Weight:
318.68
Beilstein:
3563831
EC Number:
MDL number:
UNSPSC Code:
12352300

Assay

97%

mp

179-182 °C (lit.)

SMILES string

CC(=O)O[Hg]OC(C)=O

InChI

1S/2C2H4O2.Hg/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

BRMYZIKAHFEUFJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service