Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33672

Supelco

Trifloxysulfuron sodium salt

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13F3N5NaO6S
CAS Number:
Molecular Weight:
459.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[Na+].COc1cc(OC)nc(NC(=O)[N-]S(=O)(=O)c2ncccc2OCC(F)(F)F)n1

InChI

1S/C14H14F3N5O6S.Na/c1-26-9-6-10(27-2)20-12(19-9)21-13(23)22-29(24,25)11-8(4-3-5-18-11)28-7-14(15,16)17;/h3-6H,7H2,1-2H3,(H2,19,20,21,22,23);/q;+1/p-1

InChI key

UFEIWEXHHOXPGP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Trifloxysulfuron sodium salt is a broad-spectrum sulfonylurea herbicide, used for the control of weeds in sugarcane and citrus plants.
Trifloxysulfuron sodium salt is a sulfonylurea broad-spectrum herbicide, particularly developed for weed control in sugarcane and cotton. Its mode of action involves the inhibition of acetolactate synthase enzyme activity.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of growth stage on trifloxysulfuron and glyphosate efficacy in twelve weed species of citrus groves.
Singh S and Singh M
Weed Technology, 18(4), 1031-1036 (2004)
Trifloxysulfuron-sodium: a new post emergence herbicide for use in cotton and sugarcane
The BCPC Conference Weeds 2001, 29-34 (2001)
Suitable adjuvant to maximize trifloxysulfuron efficacy and early assessment of herbicide efficacy using chlorophyll fluorescence
Singh S and Singh M
Pesticide Formulations and Delivery Systems, 1(1470), 103-103 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service