Skip to Content
MilliporeSigma
All Photos(1)

Documents

33660

Supelco

PCB No 118

analytical standard

Synonym(s):

2,3′,4,4′,5′-Pentachlorobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H5Cl5
CAS Number:
Molecular Weight:
326.43
Beilstein:
2379471
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

SMILES string

Clc1ccc(cc1Cl)-c2cc(Cl)c(Cl)cc2Cl

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service