Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33412

Supelco

2,3,4-Trichloroanisole

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5Cl3O
CAS Number:
Molecular Weight:
211.47
Beilstein:
2364316
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1ccc(Cl)c(Cl)c1Cl

InChI

1S/C7H5Cl3O/c1-11-5-3-2-4(8)6(9)7(5)10/h2-3H,1H3

InChI key

FRQUNVLMWIYOLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3,4-Trichloroanisole may be used as a reference standard for the determination of the analyte in water samples using stir bar sorptive extraction (SBSE), followed by thermal desorption-capillary gas chromatography coupled to mass spectrometry (GC/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of off-flavors in the aquatic environment by stir bar sorptive extraction--thermal desorption--capillary GC/MS/olfactometry.
Benanou D, et al.
Analytical and Bioanalytical Chemistry, 376(1), 69-77 (2003)
William J S Lockley et al.
Journal of labelled compounds & radiopharmaceuticals, 63(13), 531-552 (2020-09-05)
Catalytic dehalogenation of aromatic halides using isotopic hydrogen gas is an important strategy for labelling pharmaceuticals, biochemicals, environmental agents and so forth. To extend, improve and further understand this process, studies have been carried out on the scrambling of deuterium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service