Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33381

Supelco

Ioxynil-octanoate

PESTANAL®, analytical standard

Synonym(s):

3,5-Diiodo-4-hydroxybenzonitrile octanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17I2NO2
CAS Number:
Molecular Weight:
497.11
Beilstein:
2756640
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

52-62 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCCCCC(=O)Oc1c(I)cc(cc1I)C#N

InChI

1S/C15H17I2NO2/c1-2-3-4-5-6-7-14(19)20-15-12(16)8-11(10-18)9-13(15)17/h8-9H,2-7H2,1H3

InChI key

QBEXFUOWUYCXNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ioxynil-octanoate is a contact herbicide used to control weeds in cereals.

Application

Ioxynil-octanoate may be used as a reference standard to quantify the evaporated content of the analyte in soil samples using simple passive dosimeters, followed by liquid chromatography with diode array detection (LC-DAD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pesticides Abstracts, 14 (1981)
S C K Carlsen et al.
Chemosphere, 64(5), 787-794 (2005-12-13)
In the present study the evaporation of 10 herbicides was investigated during five field experiments, and the amount deposited per surface area was quantified inside the field using simple passive dosimeters consisting of microscope slides placed on plastic lids. On

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service