Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33211-M

Sigma-Aldrich

Ethyl acetate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

GC/GC: suitable

impurities

≤0.001% non-volatile matter
≤0.0045% free acid (as CH3COOH)
≤0.05% water (Karl Fischer)
≤0.1% ethanol (GC)
≤0.1% isobutylalcohol
≤0.1% methanol
≤0.1% methyl acetate

color

APHA: >10

refractive index

n20/D 1.3710-1.3730
n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CCOC(C)=O

suitability

passes test for reaction against H2SO4

format

neat

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl Acetate (EtOAc) is an organic compound with a characteristic sweet smell. It is commonly used as a coating solvent for paints, lacquers, and varnishes. It is also used as an extraction solvent and reagent in many pharmaceutical industries.

Application

Ethyl acetate can be used as:     
  • A reactant to synthesize cinnamyl acetate via lipase-catalyzed transesterification reaction between ethyl acetate and cinnamyl alcohol.      
  • A reaction medium to synthesize solid electrolyte Li3PS4 by the reaction between Li2S and P2S5.      
  • A solvent in the synthesis of 6-O-acetyl glycosides via selective acetylation of carbohydrates in the presence of a catalytic amount of sulfuric acid. 6-O-acetyl glycoside derivatives are applicable as precursors for the synthesis of complex oligosaccharides.      
  • A reactant to prepare geranyl acetate by transesterification of geraniol.      
  • A green solvent in solid-phase peptide synthesis.

Features and Benefits

EtOAc is an environmentally safe solvent/reagent.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service