Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

33006

Supelco

Benthiavalicarb isopropyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$361.00

$361.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$361.00

About This Item

Empirical Formula (Hill Notation):
C18H24FN3O3S
CAS Number:
Molecular Weight:
381.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$361.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

CC(C)OC(=O)N[C@@H](C(C)C)C(=O)N[C@H](C)c1nc2ccc(F)cc2s1

InChI

1S/C18H24FN3O3S/c1-9(2)15(22-18(24)25-10(3)4)16(23)20-11(5)17-21-13-7-6-12(19)8-14(13)26-17/h6-11,15H,1-5H3,(H,20,23)(H,22,24)/t11-,15?/m1/s1

InChI key

USRKFGIXLGKMKU-ZRKZCGFPSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Benthiavalicarb isopropyl is classified under the class of amino acid amide carbamate category of fungicides.[1]

Application

Benthiavalicarb isopropyl may be used as an analytical reference standard for the determination of benthiavalicarb isopropyl in:
  • Fruits and vegitables by ultra-performance liquid chromatography coupled to quadrupole-time-of-flight-mass spectrometry (UPLC-QTOF-MS) equipped with automated detection.[2]
  • Food samples[3] and human serum samples[4] by QuEChERS (quick, easy, cheap, effective, rigged, and safe) extraction LC coupled to electrospray ionization-tandem mass spectrometry (ESI-MS/MS) equipped with multiple reaction monitoring (MRM) detection.
  • Chinese cabbage and cucumber samples by QuEChERS extraction, primary secondary amine dispersive solid-phase extraction (PSA d-SPE) and LC-ESI-MS/MS with selected reaction monitoring (SRM) detection.[5]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fungicidial activity of benthiavalicarb-isopropyl against Phytophthora infestans and its controlling activity against late blight diseases.
Miyake Y, et al.
Journal of Pesticide Science, 30(4), 390-396 (2005)
Evaluation and validation of an accurate mass screening method for the analysis of pesticides in fruits and vegetables using liquid chromatography-quadrupole-time of flight-mass spectrometry with automated detection.
Lopez MG, et al.
Journal of Chromatography A, 1373(4), 40-50 (2014)
Validation and use of a fast sample preparation method and liquid chromatography-tandem mass spectrometry in analysis of ultra-trace levels of 98 organophosphorus pesticide and carbamate residues in a total diet study involving diversified food types.
Chung SWC and Chan BTP
Journal of Chromatography A, 1217(29), 4815-4824 (2010)
Validation of a Multiresidue Analysis Method for 379 Pesticides in Human Serum Using Liquid Chromatography?Tandem Mass Spectrometry.
Shin Y, et al.
Journal of Agricultural and Food Chemistry, 66(13), 3550-3560 (2018)
Mei-Ai Zhao et al.
Journal of agricultural and food chemistry, 62(47), 11449-11456 (2014-11-08)
This paper describes the comparison of five sample cleanup procedures for the determination of 238 pesticides via triple quadrupole liquid chromatography-tandem mass spectrometry (LC-MS/MS, with only 10 min of chromatographic running time) in Chinese cabbage and cucumber. Samples were extracted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service