Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32958

Supelco

DMST solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Synonym(s):

N,N-Dimethyl-N′-tolylsulfonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N2O2S
CAS Number:
Molecular Weight:
214.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CN(C)S(=O)(=O)Nc1ccc(C)cc1

InChI

1S/C9H14N2O2S/c1-8-4-6-9(7-5-8)10-14(12,13)11(2)3/h4-7,10H,1-3H3

InChI key

UDCDOJQOXWCCSD-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service