Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32871

Supelco

Cyclanilide

PESTANAL®, analytical standard

Synonym(s):

1-[(2,4-Dichlorophenyl)aminocarbonyl]-1-cyclopropanecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9Cl2NO3
CAS Number:
Molecular Weight:
274.10
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)C1(CC1)C(=O)Nc2ccc(Cl)cc2Cl

InChI

1S/C11H9Cl2NO3/c12-6-1-2-8(7(13)5-6)14-9(15)11(3-4-11)10(16)17/h1-2,5H,3-4H2,(H,14,15)(H,16,17)

InChI key

GLWWLNJJJCTFMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclanilide is typically employed as a plant growth inhibitor in cotton, either to inhibit vegetative growth (in combination with mepiquat chloride) or to promote senescence (in combination with ethephon).[1]

Application

Cyclanilide may be used as an analytical reference standard for the determination of the analyte in:
  • Infant formula[2] and milk samples[3] by ultra-performance liquid chromatography–tandem mass spectrometry (UPLC-MS/MS).
  • Cotton seed oil samples by solid-phase extraction (SPE) and high performance liquid chromatography (HPLC) with ultraviolet (UV) detection.[4]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of cyclanilide on auxin activity.
Burton JD, et al.
Journal of Plant Growth Regulation, 27(4), 342-342 (2008)
A novel method for determination of cyclanilide and its metabolite residues in cotton seed oil.
Rao TN, et al.
The experiment, 22(2), 1531-1536 (2014)
Generic and rapid determination of veterinary drug residues and other contaminants in raw milk by ultra performance liquid chromatography-tandem mass spectrometry.
Zhan J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 906(2-3), 48-57 (2012)
Multi-class method for determination of veterinary drug residues and other contaminants in infant formula by ultra performance liquid chromatography-tandem mass spectrometry.
Zhan J, et al.
Food Chemistry, 138(2-3), 827-834 (2013)
Nikolay Vasilev et al.
Scientific reports, 6, 37390-37390 (2016-11-18)
Multiple factors act simultaneously on plants to establish complex interaction networks involving nutrients, elicitors and metabolites. Metabolomics offers a better understanding of complex biological systems, but evaluating the simultaneous impact of different parameters on metabolic pathways that have many components

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service