Skip to Content
MilliporeSigma
All Photos(1)

Documents

32826

Supelco

Hydroxymethylclenbuterol

VETRANAL®, analytical standard

Synonym(s):

2-[2-(4-Amino-3,5-dichlorophenyl)-2-hydroxyethylamino]-2-methyl-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18Cl2N2O2
CAS Number:
Molecular Weight:
293.19
Beilstein:
2854376
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(CO)NCC(O)c1cc(Cl)c(N)c(Cl)c1

InChI

1S/C12H18Cl2N2O2/c1-12(2,6-17)16-5-10(18)7-3-8(13)11(15)9(14)4-7/h3-4,10,16-18H,5-6,15H2,1-2H3

InChI key

BWURCANZQUYPLR-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Blokland et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(9), 1703-1715 (2018-08-15)
Nowadays routine residue monitoring involves the analysis of many compounds from different classes, mainly in urine. In the past two decades, developments heavily focused on the use of mass spectrometers (MS) and faster and more sensitive MS detectors have reached

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service