Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32168

Supelco

Aminotriclabendazole hydrochloride

VETRANAL®, analytical standard

Synonym(s):

5-Chloro-6-(2,3-dichlorophenoxy)-1H-benzoimidazol-2-ylamine monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8Cl3N3O · HCl
CAS Number:
Molecular Weight:
365.04
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

NC1=NC(C=C2Cl)=C(N1)C=C2OC3=C(Cl)C(Cl)=CC=C3.Cl

InChI

1S/C13H8Cl3N3O.ClH/c14-6-2-1-3-10(12(6)16)20-11-5-9-8(4-7(11)15)18-13(17)19-9;/h1-5H,(H3,17,18,19);1H

InChI key

VIRKTPJWYSJBRP-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service