Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31639

Supelco

2,4-Diaminoanisole sulfate salt hydrate

analytical standard

Synonym(s):

4-Methoxy-m-phenylenediamine sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NH2)2·H2SO4·xH2O
CAS Number:
Molecular Weight:
236.25 (anhydrous basis)
Beilstein:
8397141
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

192 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

O.OS(O)(=O)=O.COc1ccc(N)cc1N

InChI

1S/C7H10N2O.H2O4S.H2O/c1-10-7-3-2-5(8)4-6(7)9;1-5(2,3)4;/h2-4H,8-9H2,1H3;(H2,1,2,3,4);1H2

InChI key

JAYRYWNGILOCLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Muta. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service