Skip to Content
MilliporeSigma
All Photos(1)

Documents

31637

Supelco

Benfuresate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16O4S
CAS Number:
Molecular Weight:
256.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCS(=O)(=O)Oc1ccc2OCC(C)(C)c2c1

InChI

1S/C12H16O4S/c1-4-17(13,14)16-9-5-6-11-10(7-9)12(2,3)8-15-11/h5-7H,4,8H2,1-3H3

InChI key

QGQSRQPXXMTJCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benfuresate is a herbicide which is specifically effective against Digitaria, Sectaria, Cyperus, Portulaca, Solanum and Gallium weeds. Benfuresate mode of action involves inhibiting fat synthesis.

Application

Benfuresate may be used as a reference standard in the determination of the analyte in water and urine samples by an analytical method based on coupling photo-induced chemiluminescence in a multicommutation continuous flow methodology.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Albert-García et al.
Talanta, 75(3), 717-724 (2008-07-01)
The present paper deals with an analytical strategy based on coupling photo-induced chemiluminescence in a multicommutation continuous-flow methodology for the determination of the herbicide benfuresate. The solenoid valve inserted as small segments of the analyte solution was sequentially alternated with
Jin-Young Lee et al.
Pesticide biochemistry and physiology, 172, 104751-104751 (2021-02-02)
Benfuresate (2,3-dihydro-3,3-dimethylbenzofuran-5-yl ethanesulphonate) is a widely used pre-emergence herbicide of the benzofurane group, which works through the inhibition of lipid synthesis. During embryonic development of zebrafish, benfuresate retards growth while causing internal changes in the body, including alteration of the
Christa Lechelt-Kunze et al.
Pest management science, 59(8), 847-856 (2003-08-15)
In order to study the mode of action of herbicides we conducted a pilot study analysing phenotype and gene expression of flufenacet- and benfuresate-treated Arabidopsis thaliana (L) Heynhoe plants. Treatments with either herbicide caused phenocopies of the known Arabidopsis mutant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service