Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31211

Supelco

Heptachlor solution

100 μg/mL in methanol, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5Cl7
CAS Number:
Molecular Weight:
373.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

ClC1C=CC2C1[C@]3(Cl)C(Cl)=C(Cl)[C@@]2(Cl)C3(Cl)Cl

InChI

1S/C10H5Cl7/c11-4-2-1-3-5(4)9(15)7(13)6(12)8(3,14)10(9,16)17/h1-5H/t3?,4?,5?,8-,9+/m1/s1

InChI key

FRCCEHPWNOQAEU-ZFBAKZBRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptachlor, a chlorinated cyclodiene, is generally used as chiral pesticide.

Application

Heptachlor may be used as an insecticide analytical standard for the determination of the analyte:
  • In groundwater samples by solid phase microextraction (SPME) method with a polyacrylate fibre, prior to detection by gas chromatography mass spectrometry in electron impact ionisation mode (GC–EI-MS).
  • In fruits and vegetables using solid phase extraction clean-up followed by gas chromatography and electron-capture detection (GC–ECD).
Heptachlor was used in determination of concentration levels of heptachlor in dry fish using gas chromatography with electron capture detector.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sources, atmospheric transport and deposition mechanism of organochlorine pesticides in soils of the Tibetan Plateau.
Chen L
The Science of the Total Environment, 577, 405-412 (2017)
Incidence of organochlorine insecticides (DDT and heptachlor) in Bangladeshi dry fish: Seasonal trends and species variability
Bhuiyan, N.I., Bhuiyan, Habibur Rahman; and Nath, K.K.
African Journal of Environmental Science and Technology (2009)
Determination of organochlorine and pyrethroid pesticides in fruit and vegetables using solid phase extraction clean-up cartridges.
Sharif Z, et al.
Journal of Chromatography A, 1127(1-2), 254-261 (2006)
Sérgio Sousa et al.
Environmental technology, 32(5-6), 673-683 (2011-09-01)
Pesticides have been responsible for strong environmental impacts, mainly due to their persistence in the environment. Removal technologies are usually combined, because degradation of organic matter is needed prior to a tertiary treatment to guarantee pesticides elimination to levels below
Syed Ali-Musstjab-Akber-Shah Eqani et al.
Bulletin of environmental contamination and toxicology, 88(3), 303-310 (2011-12-17)
This review evaluates and summarizes the results of the studies of organochlorines-contaminated water, sediment/soil and biota in Pakistan. Pattern of occurrence of each contaminant class from different study sites is followed as DDTs > Cyclodiens > HCHs > PCBs. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service