Skip to Content
MilliporeSigma
All Photos(1)

Documents

30884

Supelco

Pigment Yellow 3

analytical standard

Synonym(s):

2-[(4-Chloro-2-nitrophenyl)azo]-N-(2-chlorophenyl)-3-oxobutyramide, 2-[2-(4-Chloro-2-nitrophenyl)diazenyl]-N-(2-chlorophenyl)-3-oxobutanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12Cl2N4O4
CAS Number:
Molecular Weight:
395.20
Colour Index Number:
11710
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

O=C(C)C(C(NC1=CC=CC=C1Cl)=O)/N=N/C2=C([N+]([O-])=O)C=C(Cl)C=C2

InChI

1S/C16H12Cl2N4O4/c1-9(23)15(16(24)19-12-5-3-2-4-11(12)18)21-20-13-7-6-10(17)8-14(13)22(25)26/h2-8,15H,1H3,(H,19,24)/b21-20+

InChI key

QTSNFLIDNYOATQ-QZQOTICOSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service