Skip to Content
MilliporeSigma
All Photos(1)

Documents

305464

Sigma-Aldrich

Fast Red B tetrafluoroborate salt

Dye content 95 %

Synonym(s):

2-Methoxy-4-nitrobenzenediazonium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NO2)N2BF4
CAS Number:
Molecular Weight:
266.95
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

composition

Dye content, 95%

Quality Level

mp

122-124 °C (lit.)

λmax

375 nm

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.COc1cc(ccc1[N+]#N)[N+]([O-])=O

InChI

1S/C7H6N3O3.BF4/c1-13-7-4-5(10(11)12)2-3-6(7)9-8;2-1(3,4)5/h2-4H,1H3;/q+1;-1

InChI key

GYYABOARHFFQTR-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Gordeeva et al.
Archives of toxicology, 95(1), 253-269 (2020-09-15)
Pluripotent stem cells recapitulate in vitro the early developmental stages and are considered promising cell models for predictive developmental toxicity studies. To investigate the consistency between adverse drug effects on early development and the early stages of embryonic stem cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service