Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30417-M

Sigma-Aldrich

Phosphoric acid

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%

Synonym(s):

Orthophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein:
1921286
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor density

3.4 (vs air)

vapor pressure

2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)

Assay

≥85%

form

liquid

impurities

reducing impurities, in accordance
residual solvents, in accordance
substances precipitated by ammonia, in accordance
≤0.001% KMnO4 red. matter (as O)
≤0.001% heavy metals (as Pb)
≤0.001% insoluble matters (in water)
≤0.001% volatile acids (as CH3COOH)

bp

158 °C (lit.)

mp

~40 °C (lit.)

density

1.685 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.0002%
fluoride (F-): ≤0.0001%
nitrate (NO3-): ≤0.0003%
phosphite, hypophosphite (as H3PO3): ≤0.002%
phosphite, hypophosphite (as H3PO3): in accordance
sulfate (SO42-): ≤0.003%

cation traces

As: ≤0.00005%
Ca: ≤0.001%
Cd: ≤0.0001%
Co: ≤0.0001%
Cr: ≤0.0001%
Cu: ≤0.0001%
Fe: ≤0.0005%
K: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.00005%
Na: ≤0.002%
Ni: ≤1 mg/kg
Pb: ≤0.0001%
Sb: ≤2 mg/kg
Zn: ≤0.0002%

SMILES string

OP(O)(O)=O

suitability

passes test for appearance of solution

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

General description

Phosphoric acid (H3PO4) is one of the most important and useful mineral acids. It is used as a catalyst for various organic reactions. It is also used as an inorganic acid electrolyte for fuel cell applications.

Application

Phosphoric acid can be used as an activating agent for the activation of porous carbonaceous materials due to its ability to produce P-containing functional groups in the carbon matrix. These groups are excellent for promoting the formation of carbon micropores, which are useful for developing electrochemical energy storage devices.

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Fujitaki et al.
Biochemistry, 20(12), 3658-3664 (1981-06-09)
Phosphorus-31 nuclear magnetic resonance (31P NMR) is used to investigate acid-labile phosphorylation of histone H4. 31P NMR detects phosphorylated histidine residues in in vitro enzymatically phosphorylated H4. The source of kinase is nuclei from either regenerating rat liver or Walker-256
P Meneses et al.
Journal of lipid research, 29(5), 679-689 (1988-05-01)
The common phospholipids from biological sources were quantitated using phosphorus-31 nuclear magnetic resonance (NMR) spectroscopy in conjunction with an analytical reagent composed of two parts: 1) 2 ml of reagent chloroform in which was dissolved 0.01-100 mg of crude tissue
D S Marinier et al.
Journal of lipid research, 29(9), 1237-1243 (1988-09-01)
In order to study the lipid composition of human white matter, we have developed a 31P NMR spectroscopy method, which allows the determination and quantitation of the main phospholipids found in biological membranes. The technique is based upon the use

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service