Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29780

Sigma-Aldrich

Ethyl 2-oxocyclopentanecarboxylate

purum, ≥97.0% (GC)

Synonym(s):

2-(Ethoxycarbonyl)cyclopentanone, Ethyl cyclopentanone-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O=)C5H7CO2C2H5
CAS Number:
Molecular Weight:
156.18
Beilstein:
387787
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.452 (lit.)
n20/D 1.453

bp

102-104 °C/11 mmHg (lit.)

density

1.054 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C1CCCC1=O

InChI

1S/C8H12O3/c1-2-11-8(10)6-4-3-5-7(6)9/h6H,2-5H2,1H3

InChI key

JHZPNBKZPAWCJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phase-transfer benzylation reaction of ethyl 2-oxocyclopentanecarboxylate with benzyl bromide has been investigated in microreactor. Ethyl 2-oxocyclopentanecarboxylate participates in cobalt(II) Schiff′s base complex catalyzed oxidation of primary and secondary alcohols to aldehydes and ketones.

Application

Ethyl 2-oxocyclopentanecarboxylate was used in stereoselective synthesis of (±)-cis,cis-spiro[4.4]nonane-1,6-diol. It was also used in synthesis of tanikolide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new synthesis of tanikolide.
Zhai H, et al.
Tetrahedron Letters, 44(14), 2893-2894 (2003)
Masaharu Ueno et al.
Chemical communications (Cambridge, England), (8)(8), 936-937 (2003-05-15)
Phase-transfer alkylation in a microreactor proceeds smoothly, and the reaction has been found to be more efficient than that in a round-bottomed flask with vigorous stirring; we have observed by an optical microscope study that an interfacial area provided by
Cobalt (II) Schiff's base complex catalysed oxidation of alcohols with dioxygen in the presence of ethyl 2-oxocyclopentanecarboxylate.
Punniyamurthy T and Iqbal J.
Tetrahedron Letters, 35(23), 4007-4010 (1994)
An improved synthesis and resolution of (?)-< i> cis, cis</i>-spiro [4.4] nonane-1, 6-diol.
Nieman JA, et al.
Tetrahedron Asymmetry, 4(9), 1973-1976 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service