Skip to Content
MilliporeSigma
All Photos(1)

Documents

29565

Sigma-Aldrich

Cyclononanone

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H16(=O)
CAS Number:
Molecular Weight:
140.22
Beilstein:
1280879
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.477 (lit.)

bp

95-97 °C/18 mmHg (lit.)

mp

24-26 °C (lit.)

density

0.959 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCCCC1

InChI

1S/C9H16O/c10-9-7-5-3-1-2-4-6-8-9/h1-8H2

InChI key

BAUZLFKYYIVGPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service